Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Alcohol, (Isopentyl Alcohol), Reagents

Catalog No. 01337554
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-554 500 mL
01-337-555 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-554 Supplier Reagents Holdings Llc Supplier No. C2119500500C
Add to Cart
Add to Cart

Isoamyl Alcohol, (Isopentyl Alcohol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym Isopentyl alcohol
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Boiling Range 131°C to 132.5°C
Melting Point -117.2°C
Color Colorless
Boiling Point 131°C to 132.5°C
Melting Range -117.2°C
Packaging Glass Bottle
Quantity 500 mL
Specific Gravity 0.813
Grade Reagent
Physical Form Liquid
Chemical Name or Material Isoamyl Alcohol, (Isopentyl Alcohol)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.