Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Alcohol, MP Biomedicals™

Catalog No. p-7083120 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mL
500 mL
1 L

Routinely used in molecular biology. Isoamyl Alcohol, MP Biomedicals is one of several isomers of amyl alcohol. It is especially utilized as a reagent in the purification of DNA, the extraction of ethidium bromide from DNA solutions and various other misc

Isoamy alcohol is routinely used in molecular biology, especially in the purification of DNA. Isoamyl alcohol is also used in the extraction of ethidium bromide from DNA solutions and in the radiolabeling of RNA transcripts in nuclei that have been isolated from tissue.

  • Widely used in conjunction with phenol and chloroform (25:24:1—phenol:chloroform:isoamyl alcohol) for the removal of proteins from the nucleic acid solutions by extraction
  • Isoamyl alcohol is added to the phenol solution to help inhibit RNase activity and to help prevent the solubilization in the phenol phase of long RNA molecules with long poly(A) portions
  • It will also help in reducing the foaming during the extraction process

Isoamy alcohol has various miscellaneous uses, including:

  • HPLC analysis of various pharmaceuticals and metabolites
  • Analysis of oxidized and reduced pyridine nucleotides and adenylates in organic phenol extracts from mitochondria
  • Isolation of mRNA from a thermophilic cyanobacterium
  • Solvent for fats, resins and alkaloids
  • Manufacturing of isoamyl (amyl) compounds, isovaleric acid, mercury fulminate, pyroxylin, artificial silk, lacquers and smokeless powders
  • For dehydrating celloidin solutions
  • For determining fat in milk
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Melting Point -117.2°C (literature)
Density 0.81g/mL (water=1) at 25°C (Lit.)
Boiling Point 131°C (literature)
Refractive Index n20/D 1.406 (Lit.)
Quantity 100 mL
Solubility Information Miscible with alcohol, ether, benzene, chloroform, petroleum ether, glacial acetic acid, oils.
Vapor Density 3.04 (air=1) (Lit.)
Vapor Pressure 2.4mmHg at 20°C, 2.37mmHg at 25°C (Lit.)
Formula Weight 88.15
Percent Purity ≥99%
Physical Form Liquid
Show More Show Less
Recommended Storage Room Temperature (15–30°C).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.