Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Hexanoate 98.0+%, TCI America™
SDP

Catalog No. I0899100ML
Click to view available options
Quantity:
25 mL
100 mL

Chemical Identifiers

CAS 2198-61-0
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
MDL Number MFCD00027280
InChI Key XVSZRAWFCDHCBP-UHFFFAOYSA-N
Synonym Hexanoic Acid Isoamyl Ester, Isopentyl Hexanoate, Hexanoic Acid Isopentyl Ester
PubChem CID 16617
ChEBI CHEBI:87542
IUPAC Name 3-methylbutyl hexanoate
SMILES CCCCCC(=O)OCCC(C)C

Specifications

Color Colorless
Boiling Point 98°C
Quantity 100 mL
Formula Weight 186.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Hexanoate
EINECSNumber (2)-0782
RTECSNumber MO8389300
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.