Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl nitrite, 96%, stabilized, Thermo Scientific™

Catalog No. AC412751000
Click to view available options
:
100g; Plastic bottle packed in tin can

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 110-46-3, 497-19-8
Density 0.8700g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic Bottle in Tin Can
Molecular Formula C5H11NO2
Refractive Index 1.3840 to 1.3880 (20°C, 589nm)
Linear Formula (CH3)2CHCH2CH2ONO
Quantity 100g
MDL Number MFCD00002057
Beilstein 01, 402
Fieser 01,40; 04,270; 05,358; 06,307; 09,258
Merck Index 15, 5168
Synonym isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i
InChI Key OWFXIOWLTKNBAP-UHFFFAOYSA-N
SMILES CC(C)CCON=O
IUPAC Name 3-methylbutyl nitrite
Molecular Weight (g/mol) 117.15
Stabilizer 0.5% sodium carbonate
PubChem CID 8053
ChEBI CHEBI:2691
Formula Weight 117.15
Percent Purity ≥95%
Chemical Name or Material Isoamyl nitrite, 90%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-770-8
RTECSNumber NT0187500
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.