Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobornyl methacrylate, 85-90%, stabilized

Catalog No. p-150370
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle

CAS: 7534-94-3 | C14H22O2 | 222.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7534-94-3
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00081070
InChI Key IAXXETNIOYFMLW-UHFFFAOYNA-N
Synonym ibma, isobornyl methacrylate, methacrylic acid isobornyl ester, iso-bornyl methacrylate, isobornyl methacrylate, technical grade, isobornyl methacrylate, stabilized with mehq, exo-1,7,7-trimethylbicyclo 2.2.1 hept-, 2-yl methacrylate, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate
PubChem CID 71311141
IUPAC Name [(1R,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C

Specifications

Melting Point -60.0°C
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 245.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 85% min. (GC)
Packaging Glass bottle
Refractive Index 1.4760 to 1.4780
Quantity 250 g
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble
Stabilizer 100 to 200ppm MEHQ
Formula Weight 222.33
Percent Purity 85-90%
Physical Form Liquid
Chemical Name or Material Isobornyl methacrylate, 85-90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 231-403-1
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.