Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutanol, HPLC Grade, 99+%

Catalog No. AA22908K7
Click to view available options
Quantity:
1 L
4 L
4 x 1 L

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05% | CAS: 78-83-1 | C4H10O | 74.123 g/mol

Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is used as a mobile phase in High Performance Liquid Chromatography. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Melting Point -108°C
Color Colorless
Density 0.802 g/cm3
Boiling Point 107°C to 108°C
Flash Point 28°C (82°F)
Ignition Point 390°C (734°F)
Odor Sweet
Assay Percent Range >99%
Refractive Index 1.396
Quantity 4 L
UN Number 1212
Beilstein 1730878
Solubility Information Miscible with alcohol and diethyl ether. Partially miscible with water.
Vapor Pressure 4-40 hPa (3-30mm Hg) at 20°C
Formula Weight 74.12
Grade HPLC
Physical Form Liquid
Chemical Name or Material Isobutanol
Show More Show Less
Hazard Category H226-H315-H318-H335-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3, Packing Group: III
EINECSNumber 201-148-0
RTECSNumber NP9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.