Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Acetate 99.0+%, TCI America™
SDP

Catalog No. p-7116673
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 110-19-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008932
InChI Key GJRQTCIYDGXPES-UHFFFAOYSA-N
Synonym isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
PubChem CID 8038
ChEBI CHEBI:50569
IUPAC Name 2-methylpropyl acetate
SMILES CC(C)COC(C)=O

Specifications

Color Colorless
Boiling Point 117°C
Quantity 25 mL
UN Number 1213
Formula Weight 116.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Acetate
EINECSNumber (2)-0731
RTECSNumber AI4025000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.