Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isobutylamine, 99%
Click to view available options
Quantity:
250 mL
1000 mL
Description
Isobutylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isobutylamine is used as pharmaceutical intermediate.
Solubility
Soluble in water (1000 mg/ml at 25°C), ether, alcohol, acetone and benzene.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Isobutylamine is used as pharmaceutical intermediate.
Solubility
Soluble in water (1000 mg/ml at 25°C), ether, alcohol, acetone and benzene.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications
Specifications
| Melting Point | °C to 85°C |
| Density | 0.736 g/mL |
| Boiling Point | 67°C to 69°C |
| Flash Point | -9°C (15°F) |
| Assay Percent Range | 99% |
| Refractive Index | 1.397 |
| Quantity | 250 mL |
| UN Number | UN1214 |
| Beilstein | 385626 |
| Sensitivity | Air Sensitive |
| Show More |
Safety and Handling
| Hazard Category | H225-H301-H314-H335 |
| Hazard Statement | GHS H Statement H225-H301-H314-H318 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYLAMINE |
| EINECSNumber | 201-145-4 |
| RTECSNumber | NP9900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction