Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutylene Sulfide 98.0+%, TCI America™
SDP

Catalog No. I046125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I046125ML 25 mL
I04615ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I046125ML Supplier TCI America Supplier No. I046125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3772-13-2
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.17
MDL Number MFCD00191662
InChI Key HGJOFJDIHKHKAU-UHFFFAOYSA-N
Synonym 2,2-Dimethylethylene Sulfide, 2,2-Dimethylthiirane, 2-Methylpropylene Sulfide
PubChem CID 138050
IUPAC Name 2,2-dimethylthiirane
SMILES CC1(C)CS1

Specifications

Color Yellow
Boiling Point 86°C
Quantity 25 mL
UN Number 1993
Formula Weight 88.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutylene Sulfide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.