Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isobutyllithium, 1.7M solution in heptane, AcroSeal™
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Yellow |
Linear Formula | (CH3)2CHCH2Li |
Density | 0.7000g/mL |
Solubility Information | Solubility in water: vigorous reaction. |
Formula Weight | 64.06 |
Percent Purity | 15 to 17% (as iso-BuLi) |
Packaging | AcroSeal™ Glass Bottle |
Specific Gravity | 0.7 |
Quantity | 800 mL |
Physical Form | Liquid |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement May cause drowsiness or dizziness. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Catches fire spontaneously if exposed to air. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water. |
Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction