Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyraldehyde, 99+%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
78-84-2
C4H8O
72.11
MFCD00006980
AMIMRNSIRUDHCM-UHFFFAOYSA-N
isobutyraldehyde,isobutanal,isobutylaldehyde,propanal, 2-methyl,2-methylpropionaldehyde,isobutyric aldehyde,valine aldehyde,isopropylaldehyde,2-methyl-1-propanal,isobutaldehyde
6561
CHEBI:48943
2-methylpropanal
CC(C)C=O
78-84-2
C4H8O
72.11
MFCD00006980
AMIMRNSIRUDHCM-UHFFFAOYSA-N
isobutyraldehyde,isobutanal,isobutylaldehyde,propanal, 2-methyl,2-methylpropionaldehyde,isobutyric aldehyde,valine aldehyde,isopropylaldehyde,2-methyl-1-propanal,isobutaldehyde
6561
CHEBI:48943
2-methylpropanal
CC(C)C=O
Viscosity 0.45 mPa.s (20°C)
Density 0.7900g/mL
Refractive Index 1.3710 to 1.3740
Infrared Spectrum Authentic
Melting Point -65.0°C
Specific Gravity 0.79
Quantity 1L
Packaging Glass bottle
Flash Point −24°C
Color Undesignated
Boiling Point 63.0°C
Assay Percent Range 99+%
Linear Formula (CH3)2CHCHO
Beilstein 01, 671
Merck Index 15, 5199
Solubility Information Solubility in water: 75g/L (20°C). Other solubilities: miscible with ethanol, ether, carbon disulfide,, acetone, benzene, toluene and chloroform
Formula Weight 72.11
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material Isobutyraldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger