Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyraldehyde, 99+%

Catalog No. AC122510025
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

CAS: 78-84-2 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-84-2
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00006980
InChI Key AMIMRNSIRUDHCM-UHFFFAOYSA-N
Synonym isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde
PubChem CID 6561
ChEBI CHEBI:48943
IUPAC Name 2-methylpropanal
SMILES CC(C)C=O

Specifications

Melting Point -65.0°C
Density 0.7900g/mL
Boiling Point 63.0°C
Flash Point −24°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3710 to 1.3740
Linear Formula (CH3)2CHCHO
Quantity 2.5 L
Beilstein 01, 671
Specific Gravity 0.79
Merck Index 15, 5199
Solubility Information Solubility in water: 75g/L (20°C). Other solubilities: miscible with ethanol, ether, carbon disulfide,, acetone, benzene, toluene and chloroform
Viscosity 0.45 mPa.s (20°C)
Formula Weight 72.11
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Isobutyraldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond conta
EINECSNumber 201-149-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.