Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyraldehyde Diethyl Acetal 96.0+%, TCI America™
SDP

Catalog No. I028725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I028725ML 25 mL
I02875ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I028725ML Supplier TCI America Supplier No. I028725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1741-41-9
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00059384
InChI Key KZDFOVZPOBSHDH-UHFFFAOYSA-N
Synonym 1,1-Diethoxyisobutane
PubChem CID 519415
IUPAC Name 1,1-diethoxy-2-methylpropane
SMILES CCOC(C(C)C)OCC

Specifications

Color Colorless
Boiling Point 138°C
Quantity 25 mL
UN Number 1993
Formula Weight 146.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyraldehyde Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.