Learn More
Isobutyramide, 99%
Description
Isobutyramide plays an important role in chemical grafting of human serum albumin during the synthesis of sequentially assembled protein capsules. It is used in pharmaceutical manufacturing and in cellular differentiation. It finds application in the transcription of human gamma-globin gene and murine embryonic epsilon(y)-globin gene. Further, it is in the treatment of beta-thalassemia and sickle cell disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water.
Notes
Incompatible with acids, bases, strong oxidizing agents and strong reducing agents.

Specifications
Specifications
Melting Point | 127°C to 129°C |
Density | 1.013 |
Boiling Point | 216°C to 220°C |
Quantity | 100 g |
Solubility Information | Soluble in water. |
Formula Weight | 87.12 |
Percent Purity | 99% |
Chemical Name or Material | Isobutyramide |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 209-265-9 |
RTECSNumber | TX1475000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.