Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyric Acid, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
1kg
Packaging:
Glass bottle
79-31-2
C4H8O2
88.106
MFCD00002658
KQNPFQTWMSNSAP-UHFFFAOYSA-N
isobutyric acid,isobutanoic acid,2-methylpropionic acid,propanoic acid, 2-methyl,dimethylacetic acid,isopropylformic acid,iso-butyric acid,isobutyrate,cenex rp b2,acetic acid, dimethyl
6590
CHEBI:16135
2-methylpropanoic acid
CC(C)C(=O)O
79-31-2
C4H8O2
88.106
MFCD00002658
KQNPFQTWMSNSAP-UHFFFAOYSA-N
isobutyric acid,isobutanoic acid,2-methylpropionic acid,propanoic acid, 2-methyl,dimethylacetic acid,isopropylformic acid,iso-butyric acid,isobutyrate,cenex rp b2,acetic acid, dimethyl
6590
CHEBI:16135
2-methylpropanoic acid
CC(C)C(=O)O
Acylation reagent
Assay Percent Range 99+%
Linear Formula (CH3)2CHCO2H
Beilstein 02, 288
Merck Index 15, 5200
Density 0.9500g/mL
Formula Weight 88.11
Physical Form Liquid
Percent Purity ≥99%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3920 to 1.3940
Specific Gravity 0.95
Solubility Solubility in water: 210g/L (20°C). Other solubilities: miscible with alcohol, chloroform and ether
Melting Point -47.0°C
Color Colorless
Boiling Point 153.0°C to 154.0°C
Quantity 1kg
Flash Point 59°C
Chemical Name or Material Isobutyric acid, 99+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-195-7

RTECSNumber : NQ4375000

TSCA : TSCA