Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyronitrile 99%, ACROS Organics™

Click to view available options
Quantity:
1L
Packaging:
Glass bottle
78-82-0
C4H7N
69.11
MFCD00001873
LRDFRRGEGBBSRN-UHFFFAOYSA-N
isobutyronitrile,isopropyl cyanide,propanenitrile, 2-methyl,2-methylpropionitrile,isopropylnitrile,2-cyanopropane,dimethylacetonitrile,isopropyl nitrile,1-cyano-1-methylethane,isopropylkyanid
6559
CHEBI:28638
2-methylpropanenitrile
CC(C)C#N
78-82-0
C4H7N
69.11
MFCD00001873
LRDFRRGEGBBSRN-UHFFFAOYSA-N
isobutyronitrile,isopropyl cyanide,propanenitrile, 2-methyl,2-methylpropionitrile,isopropylnitrile,2-cyanopropane,dimethylacetonitrile,isopropyl nitrile,1-cyano-1-methylethane,isopropylkyanid
6559
CHEBI:28638
2-methylpropanenitrile
CC(C)C#N
Melting Point -72°C
Boiling Point 107°C to 108°C
Flash Point 8°C
Specific Gravity 0.76
Refractive Index 1.3720 to 1.374
Density 0.7600g/mL
Quantity 1L
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula (CH3)2CHCN
Beilstein 02, 294
Merck Index 15, 5201
Solubility Information Solubility in water: 35g/L (20 c). Other solubilities: soluble in alcohol and ether
Formula Weight 69.11
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Isobutyronitrile, 99%
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Fatal if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-147-5

RTECSNumber : TZ4900000

TSCA : TSCA