Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyronitrile, 99%

Catalog No. AAL14470AK
Click to view available options
Quantity:
250 mL
1000 mL

CAS: 78-82-0 | C4H7N | 69.107 g/mol

Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

Solubility
Soluble in water 35g/L (20°C).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-82-0
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00001873
InChI Key LRDFRRGEGBBSRN-UHFFFAOYSA-N
Synonym isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid
PubChem CID 6559
ChEBI CHEBI:28638
IUPAC Name 2-methylpropanenitrile
SMILES CC(C)C#N

Specifications

Melting Point -72°C
Density 0.77
Boiling Point 103°C to 104°C
Flash Point 8°C (46°F)
Refractive Index 1.374
Linear Formula (CH3)2CHCN
Quantity 250 mL
UN Number UN2284
Beilstein 1340512
Merck Index 14,5156
Solubility Information Soluble in water 35g/L (20°C).
Formula Weight 69.11
Percent Purity 99%
Chemical Name or Material Isobutyronitrile
Show More Show Less
Hazard Category H225-H300+H330-H311-H371
Hazard Statement GHS H Statement
H225-H301-H311-H330
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P309+P311-P310-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYRONITRILE
EINECSNumber 201-147-5
RTECSNumber TZ4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.