Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISOC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598681
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598681 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598681 Supplier Invitrogen™ Supplier No. PA598681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ISOC1 Polyclonal Antibody for ELISA, IHC (P), Western Blot

ISOC1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include catalytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISOC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ISOC1
Gene Accession No. Q96CN7
Gene Alias 2610034N03Rik; CGI-111; Down-regulated in nephrectomized rat kidney #1; down-regulated in nephrectomized rat kidney protein 1; DR-NR#1; DR-NR1; isoc1; isochorismatase domain containing 1; isochorismatase domain-containing protein 1; isochorismatase domain-containing protein 1; LOW QUALITY PROTEIN: isochorismatase domain-containing protein 1; RGD1307632; zgc:153699
Gene Symbols ISOC1
Host Species Rabbit
Immunogen Recombinant Human Isochorismatase domain-containing protein 1 protein (155-298AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51015
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.