Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ISOC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560256
Description
Immunogen sequence: GLQVHVVVDA CSSRSQVDRL VALARMRQSG AFLSTSEGLI LQLVGDAVHP QFKEIQKLIK EPAPDSGLLG LFQGQNSLLH Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 90%.
ISOC2 gene ontology annotations related to this gene include protein destabilization.
Specifications
ISOC2 | |
Polyclonal | |
Unconjugated | |
ISOC2 | |
0610042E07Rik; AI553388; Isoc2; Isoc2a; Isoc2b; isochorismatase domain containing 2; isochorismatase domain containing 2a; isochorismatase domain containing 2b; isochorismatase domain-containing protein 2; isochorismatase domain-containing protein 2, mitochondrial; Isochorismatase domain-containing protein 2A; isochorismatase domain-containing protein 2A, mitochondrial; Isochorismatase domain-containing protein 2B; isochorismatase domain-containing protein 2B, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79763 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96AB3 | |
ISOC2 | |
Recombinant protein corresponding to Human ISOC2. Recombinant protein control fragment (Product #RP-98849). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction