Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isocitrate Dehydrogenase 1/IDH1 Antibody (IDH1/6806R), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314080F
This item is not returnable.
View return policy
Description
Isocitrate Dehydrogenase 1/IDH1 Monoclonal antibody specifically detects Isocitrate Dehydrogenase 1/IDH1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Isocitrate Dehydrogenase 1/IDH1 | |
| Monoclonal | |
| FITC | |
| Cytosolic NADP-isocitrate dehydrogenase, EC 1.1.1.42, IDCD, IDH, IDP, IDPC, isocitrate dehydrogenase [NADP] cytoplasmic, isocitrate dehydrogenase 1 (NADP+), soluble, NADP(+)-specific ICDH, NADP-dependent isocitrate dehydrogenase, cytosolic, NADP-dependent isocitrate dehydrogenase, peroxisomal, Oxalosuccinate decarboxylase, PICD | |
| Recombinant fragment of human Isocitrate Dehydrogenase 1/IDH1 protein (around aa 281-414) (exact sequence is proprietary) (Uniprot: O75874 ) | |
| 0.1 mL | |
| Stem Cell Markers | |
| 3417 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| IDH1/6806R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction