Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isocitrate Dehydrogenase 1/IDH1 Antibody (IDH1.R132H/7328R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32063320UG
Description
Isocitrate Dehydrogenase 1/IDH1 Monoclonal antibody specifically detects Isocitrate Dehydrogenase 1/IDH1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Isocitrate Dehydrogenase 1/IDH1 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
Cytosolic NADP-isocitrate dehydrogenase, EC 1.1.1.42, IDCD, IDH, IDP, IDPC, isocitrate dehydrogenase [NADP] cytoplasmic, isocitrate dehydrogenase 1 (NADP+), soluble, NADP(+)-specific ICDH, NADP-dependent isocitrate dehydrogenase, cytosolic, NADP-dependent isocitrate dehydrogenase, peroxisomal, Oxalosuccinate decarboxylase, PICD | |
Recombinant fragment (around aa 1-200) of human Isocitrate Dehydrogenase 1/IDH1 (exact sequence is proprietary) | |
20 μg | |
Stem Cell Markers | |
3417 | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
IDH1.R132H/7328R | |
Unconjugated | |
1mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction