Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoconazole nitrate, 99%, Thermo Scientific™

Catalog No. p-7077746
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 24168-96-5
Molecular Formula C18H15Cl4N3O4
Molecular Weight (g/mol) 479.14
MDL Number MFCD01721647
InChI Key NNGQLSIGRSTLLU-UHFFFAOYNA-N
Synonym isoconazole nitrate, adestan g100, travogyn, fazol, isoconazole; nitric acid, isoconazole nitrate jan, 1-2,4-dichloro-beta-2,6-dichlorobenzyloxy phenethyl imidazole nitrate, 1-2-2,6-dichlorobenzyl oxy-2-2,4-dichlorophenyl ethyl imidazole nitrate
PubChem CID 159968
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
SMILES O[N+]([O-])=O.ClC1=CC=C(C(CN2C=CN=C2)OCC2=C(Cl)C=CC=C2Cl)C(Cl)=C1

Specifications

Melting Point 181.0°C to 183.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 479.14
Percent Purity 99%
Chemical Name or Material Isoconazole nitrate
EINECSNumber 246-051-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.