Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isodehydroacetic acid, 98%, Thermo Scientific™

Catalog No. AC351520050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-65-9
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00023864
InChI Key QOADIMYPCZMZSG-UHFFFAOYSA-N
Synonym 4,6-dimethyl-2-oxo-2h-pyran-5-carboxylic acid, isodehydroacetic acid, isodehydracetic acid, 4,6-dimethyl-5-formylpyran-2-one, 2h-pyran-5-carboxylic acid, 4,6-dimethyl-2-oxo, 4,6-dimethyl-2-oxopyran-5-carboxylic acid, 4,6-dimethyl-5-carboxy-2-oxo-2h-pyran, 2h-pyran-5-carboxylic acid,6-dimethyl-2-oxo, isodehydracetsaure, 2,4-dimethyl-6-oxo-pyran-3-carboxylic acid
PubChem CID 95775
IUPAC Name 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylic acid
SMILES CC1=C(C(O)=O)C(C)=CC(=O)O1

Specifications

Melting Point 154°C to 159°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Isodehydroacetic acid, 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.