Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isohexane, for HPLC, contains <5% n-Hexane

Catalog No. AC383810010
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy

Catalog No. AC383810010

Supplier: Thermo Scientific Chemicals 383810010

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 73513-42-5 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73513-42-5
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009406
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
IUPAC Name 2-methylpentane
SMILES CCCC(C)C

Specifications

Melting Point -154.0°C
Density 0.6600g/mL
Boiling Point 60.0°C
Flash Point −23°C
Refractive Index 1.3700 to 1.3720
Linear Formula CH3CH2CH2CH(CH3)2
Quantity 1 L
Specific Gravity 0.66
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 86.18
Grade HPLC
Chemical Name or Material Isohexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.