Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isohexane, for HPLC, contains <5% n-Hexane, Acros Organics

Click to view available options
Quantity:
1L
2.5L
73513-42-5
C6H14
86.18
MFCD00009406
AFABGHUZZDYHJO-UHFFFAOYSA-N
isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane
7892
2-methylpentane
CCCC(C)C
73513-42-5
C6H14
86.18
MFCD00009406
AFABGHUZZDYHJO-UHFFFAOYSA-N
isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane
7892
2-methylpentane
CCCC(C)C
Melting Point -154.0°C
Boiling Point 60.0°C
Refractive Index 1.3700 to 1.3720
Density 0.6600g/mL
Quantity 2.5L
Flash Point −23°C
Specific Gravity 0.66
Linear Formula CH3CH2CH2CH(CH3)2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 86.18
Grade HPLC
Chemical Name or Material Isohexane
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Danger