Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoindoline, 97%

Catalog No. p-4089478
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 496-12-8 | C8H9N | 119.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-12-8
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.17
MDL Number MFCD00605324
InChI Key GWVMLCQWXVFZCN-UHFFFAOYSA-N
Synonym isoindoline, 1h-isoindole, 2,3-dihydro, iso-indoline, 2-azaindan, 1,2-dihydroisoindole, 1,3-dihydroisoindole, dihydroisoindole, pubchem16248, acmc-1am9e
PubChem CID 422478
IUPAC Name 2,3-dihydro-1H-isoindole
SMILES C1C2=CC=CC=C2CN1

Specifications

Melting Point 17°C
Color White
Density 1.0500g/mL
Boiling Point 221°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5670 to 1.5700 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.05
Formula Weight 119.17
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material Isoindoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.