Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mystaire™ Isola™ Polypropylene Base Cabinet

Polypropylene constructed cabinet ideal for storage of corrosive chemicals. For use with the Isola™ filtered workstations.
Supplier: Mystaire™ MYPBC48
Description
- 36 inches high
- Variety of shelving options
- Polypropylene construction
- Color: White
Specifications
Isola Filtered Workstation | |
28 in. | |
48 in. | |
711.2 x 1219.2 x 914.4 mm (28 x 48 x 36 in.) | |
711.2 mm | |
Base Cabinet | |
White |
36 in. | |
28 in. | |
914.4 mm | |
711.2 mm | |
1219.2 mm | |
Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction