Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott™ Isolator™ Microbial Tubes
Shop All Abbott Rapid Diagnostics Products
Click to view available options
Tube Size:
Adult
Pediatric
Volume (Metric):
1.5 mL
10 mL
Description
Benefits:
- Blood culture system for the collection and isolation of microorganisms from the blood. The ISOLATOR™ tube is used to concentrate and deliver microbial organisms to conventional growth media for purposes of isolation, identification and susceptibility testing.
- When compared to conventional broth systems, lysis centrifugation can reduce the time to isolation by 24 hours
Specifications
Specifications
| Product Type | Microbial Tube |
| For Use With (Application) | For collection and isolation of microorganisms from blood |
| For Use With (Equipment) | ISOSTAT™ System Press and Rack |
| Quantity | 25 Pk. |
| Storage Requirements | 2°C to 40°C |
| Tube Size | Adult |
| Volume (Metric) | 10 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction