Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ Isomenthone, ≥90% (GC), MilliporeSigma™ Supelco™

Catalog No. 111002050 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
100 mg

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

TRUSTED_SUSTAINABILITY

Specifications

CAS 491-07-6
Product Line phyproof
Linear Formula C10H18 O
MDL Number MFCD20760181
Synonym cis-2-Isopropyl-5-methylcyclohexanone; cis-p-Menthan 3-one; cis-Menthone
Type Reference Substance
Quantity 100 mg
Formula Weight 154.25
Percent Purity ≥90% (GC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.