Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isoniazid, ≥99% (TLC), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
5 g
50 g
250 g
Description
Pharmaceutical (small molecule)

Specifications
Specifications
Packaging | Poly Bottle |
CAS | 54-85-3 |
Physical Form | Neat |
Linear Formula | C6H7N3 O |
MDL Number | MFCD00006426 |
Melting Point | 171°C to 173°C |
Synonym | 4-Pyridinecarboxylic acid hydrazide; INH; Isonicotinic acid hydrazide; Isonicotinic hydrazide |
Type | Analytical Standard |
Quantity | 50 g |
Formula Weight | 137.14 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction