Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isonicotinic acid hydrazide, 99%

Catalog No. AC122605000
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle

CAS: 54-85-3 | C6H7N3O | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-85-3
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006426
InChI Key QRXWMOHMRWLFEY-UHFFFAOYSA-N
Synonym isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide
PubChem CID 3767
ChEBI CHEBI:6030
IUPAC Name pyridine-4-carbohydrazide
SMILES C1=CN=CC=C1C(=O)NN

Specifications

Melting Point 171°C to 173°C
Color Colorless or White
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Beilstein 22, I, 504
Merck Index 15, 5232
Solubility Information Solubility in water: 14% in water (25 c). Other solubilities: 2% in alcohol (25 c),0.1% in chloroform
Formula Weight 137.14
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Isonicotinic acid hydrazide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 200-214-6
RTECSNumber NS1751850
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.