Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isonicotinoyl chloride hydrochloride, 97%

Catalog No. AC211681000
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Plastic bottle

CAS: 39178-35-3 | C6H5Cl2NO | 178.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39178-35-3
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.01
MDL Number MFCD00012830
InChI Key BNTRVUUJBGBGLZ-UHFFFAOYSA-N
Synonym isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl
PubChem CID 12262826
IUPAC Name pyridine-4-carbonyl chloride;hydrochloride
SMILES [H+].[Cl-].ClC(=O)C1=CC=NC=C1

Specifications

Melting Point 153.0°C to 161.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Plastic bottle
Quantity 100 g
Beilstein 22, 46
Formula Weight 178.02
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Isonicotinoyl chloride hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 254-331-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.