Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopentyl acetate, 99%, cont. ca 10% other isomers, Thermo Scientific Chemicals

Catalog Number AAB21618AP
Click to view available options
Quantity:
100 mL
500 mL
123-92-2
C7H14O2
130.19
MFCD00008946
MLFHJEHSLIIPHL-UHFFFAOYSA-N
isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate
31276
CHEBI:31725
3-methylbutyl acetate
CC(C)CCOC(C)=O
123-92-2
C7H14O2
130.19
MFCD00008946
MLFHJEHSLIIPHL-UHFFFAOYSA-N
isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate
31276
CHEBI:31725
3-methylbutyl acetate
CC(C)CCOC(C)=O

It is used as a masking agent, perfuming agent, and solvent in cosmetics industry. Mainly used in flavors and because of its intensive fruity taste especially for fruit flavors such as banana, strawberry, pear, apple and cherry flavors. Traces are used in certain sweet spicy types such as vanilla flavors. Occasionally used in perfumery when a fresh-fruity note is demanded. The major application is the use in nail polish removers and nail polish as a solvent. Isopentyl acetate is a widely employed ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a masking agent, perfuming agent, and solvent in cosmetics industry. Mainly used in flavors and because of its intensive fruity taste especially for fruit flavors such as banana, strawberry, pear, apple and cherry flavors. Traces are used in certain sweet spicy types such as vanilla flavors. Occasionally used in perfumery when a fresh-fruity note is demanded. The major application is the use in nail polish removers and nail polish as a solvent. Isopentyl acetate is a widely employed ester.

Solubility
Soluble in alcohol, fixed oils, kerosene and paraffin oil. Insoluble in water, glycerin and propylene glycol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point -78°C
Density 0.876
Boiling Point 142°C
Flash Point 25°C (77°F)
Odor Fruit-like
Refractive Index 1.4
Quantity 500 mL
UN Number UN1104
Merck Index 14,5111
Solubility Information Soluble in alcohol,fixed oils,kerosene and paraffin oil. Insoluble in water,glycerin and propylene glycol.
Formula Weight 130.19
Percent Purity 99%
Assay cont. ≈10% other isomers
Chemical Name or Material Isopentyl acetate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMYL ACETATES
EINECSNumber 204-662-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit