Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophthalic Dihydrazide 95.0+%, TCI America™
SDP

Catalog No. I0257100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0257100G 100 g
I025725G 25 g
I0257500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0257100G Supplier TCI America Supplier No. I0257100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2760-98-7
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.194
MDL Number MFCD00025117
InChI Key UTTHLMXOSUFZCQ-UHFFFAOYSA-N
Synonym isophthalic dihydrazide, isophthalohydrazide, isophthalic acid dihydrazide, isophthalyl dihydrazide, 1,3-benzenedicarboxylic acid, dihydrazide, isophthalic hydrazide, isophthalic acid hydrazide, isophthalic acid, dihydrazide, 1,3-phenylenebiscarbohydrazide, unii-r51j8kl2je
PubChem CID 72700
IUPAC Name benzene-1,3-dicarbohydrazide
SMILES C1=CC(=CC(=C1)C(=O)NN)C(=O)NN

Specifications

Melting Point 227°C
Color White
Quantity 100 g
Formula Weight 194.19
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isophthalic Dihydrazide
EINECSNumber (3)-1336
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.