Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophthalonitrile, 97+%

Catalog No. p-7027273
Click to view available options
Quantity:
250 g
1000 g

CAS: 626-17-5 | C8H4N2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-17-5
Molecular Formula C8H4N2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00001795
InChI Key LAQPNDIUHRHNCV-UHFFFAOYSA-N
Synonym isophthalonitrile, 1,3-dicyanobenzene, isophthalodinitrile, 1,3-benzenedicarbonitrile, m-dicyanobenzene, 3-cyanobenzonitrile, m-benzenedinitrile, isoftalodinitril, 1,3-benzodinitrile, m-phthalodinitrile
PubChem CID 12276
ChEBI CHEBI:38218
IUPAC Name benzene-1,3-dicarbonitrile
SMILES N#CC1=CC(=CC=C1)C#N

Specifications

Melting Point 159°C to 163°C
Density 1.3
Boiling Point 288°C
Flash Point >150°C (302°F)
Odor Almond-like
Quantity 250 g
Beilstein 1072209
Formula Weight 128.13
Percent Purity 98%
Chemical Name or Material Isophthalonitrile
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-933-7
RTECSNumber CZ1900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.