Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophthaloyl dichloride, 98%

Catalog No. AC122665000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 99-63-8 | C8H4Cl2O2 | 203.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-63-8
Molecular Formula C8H4Cl2O2
Molecular Weight (g/mol) 203.02
InChI Key FDQSRULYDNDXQB-UHFFFAOYSA-N
Synonym isophthaloyl chloride, isophthaloyl dichloride, 1,3-benzenedicarbonyl dichloride, m-phthaloyl chloride, isophthalic chloride, isophthalic acid chloride, isophthalic acid dichloride, isothaloyl chloride, isophthalyl chloride, 1,3-benzenedicarbonyl chloride
PubChem CID 7451
IUPAC Name benzene-1,3-dicarbonyl chloride
SMILES C1=CC(=CC(=C1)C(=O)Cl)C(=O)Cl

Specifications

Melting Point 43.0°C to 45.0°C
Color White
Boiling Point 276.0°C
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H4(COCl)2
Quantity 500 g
Beilstein 09,IV,3295
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol,soluble in ether and other organic solvents
Formula Weight 203.02
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material Isophthaloyl dichloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
EINECSNumber 202-774-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.