Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophytol, 95%, Thermo Scientific™

Catalog No. AC352870050
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-32-8
Molecular Formula C20H40O
Molecular Weight (g/mol) 296.539
MDL Number MFCD00048380
InChI Key KEVYVLWNCKMXJX-UHFFFAOYSA-N
Synonym isophytol, 3,7,11,15-tetramethyl-1-hexadecen-3-ol, 1-hexadecen-3-ol, 3,7,11,15-tetramethyl, 2,6,10,14-tetramethylhexadec-15-en-14-ol, 2,6,10-trimethyl-14-vinylpentadecan-14-ol, 1-hexadecene-3-ol, 3,7,11,15-tetramethyl, 1-phyten-3-ol, iso-phytol, dsstox_cid_5474
PubChem CID 10453
IUPAC Name 3,7,11,15-tetramethylhexadec-1-en-3-ol
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)(C=C)O

Specifications

Color Yellow
Density 0.851g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Refractive Index 1.4550 to 1.4590 (20°C, 589nm)
Quantity 5 g
Merck Index 15, 5244
Formula Weight 296.54
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material Isophytol, 95%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.