Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Isoprenaline hemisulfate

Catalog No. AC462110050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462110050 5 g
AC462110250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462110050 Supplier Thermo Scientific Chemicals Supplier No. 462110050
Add to Cart

CAS: 299-95-6 | C11H17NO3·1/2H2O4S | 520.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-95-6
Molecular Formula C11H17NO3·1/2H2O4S
Molecular Weight (g/mol) 520.59
InChI Key ZOLBALGTFCCTJF-UHFFFAOYSA-N
Synonym isoprenaline sulphate, isoprenaline sulfate, aleudrin, novodrine, aleudrine sulfate, isoproterenol sulfate, dl-isoprenaline sulfate, dl-isoproterenol sulfate, d,l-isoproterenol sulfate, +--isoprenaline sulfate
PubChem CID 8239
IUPAC Name 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;sulfuric acid
SMILES CC(C)NCC(C1=CC(=C(C=C1)O)O)O.CC(C)NCC(C1=CC(=C(C=C1)O)O)O.OS(=O)(=O)O

Specifications

Melting Point 83.0°C
Quantity 5 g
Formula Weight 520.59
Chemical Name or Material Isoprenaline hemisulfate
EINECSNumber 206-085-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.