Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol

Catalog No. p-7046502
Click to view available options
Quantity:
100 mL
500 mL

CAS: 78-79-5 | C5H8 | 68.119 g/mol

Isoprene is used in to prepare synthetic version of natural rubber by polymerization using Ziegler catalysts. It is a basic building block for terpenes especially carotenes, terpenoids, retinol, vitamin-A and tocopherol (Vitamin-E). It finds applications in medical equipments, toys, baby bottle nipples, shoe soles, elastic films and threads for golf balls. Copolymer prepares in association with styrene is used in pressure sensitive adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoprene is used in to prepare synthetic version of natural rubber by polymerization using Ziegler catalysts. It is a basic building block for terpenes especially carotenes, terpenoids, retinol, vitamin-A and tocopherol (Vitamin-E). It finds applications in medical equipments, toys, baby bottle nipples, shoe soles, elastic films and threads for golf balls. Copolymer prepares in association with styrene is used in pressure sensitive adhesives.

Solubility
Miscible with ethanol, ethyl ether, alcohol, acetone and benzene. Immiscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases, ammonia, alkali metals, alcohols and oxygen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-79-5
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00008600
InChI Key RRHGJUQNOFWUDK-UHFFFAOYSA-N
Synonym isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren
PubChem CID 6557
ChEBI CHEBI:35194
IUPAC Name 2-methylbuta-1,3-diene
SMILES CC(=C)C=C

Specifications

Melting Point -146°C
Density 0.681
Boiling Point 33°C to 36°C
Flash Point −54°C (−65°F)
Refractive Index 1.422
Quantity 100 mL
UN Number UN1218
Beilstein 969158
Merck Index 14,5201
Solubility Information Miscible with ethanol,ethyl ether,alcohol,acetone and benzene. Immiscible with water.
Formula Weight 68.12
Percent Purity 99%
Chemical Name or Material Isoprene, stabilized with ≈0.02% 4-tert-butylcatechol
Show More Show Less
Hazard Category H224-H341-H350
Hazard Statement GHS H Statement
H224-H350-H341
Extremely flammable liquid and vapor.
May cause cancer.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P281-P303+P361+P353-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ISOPRENE, STABILIZED
EINECSNumber 201-143-3
RTECSNumber NT4037000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.