Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, 70% (v/v), Ricca Chemical
SDP

Catalog No. 421032
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
Safety-coated Glass Bottle

Chemical Identifiers

CAS 67-63-0, 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) Mixture
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Min % 29.0, 69.0
CAS Max % 31.0, 71.0
Appearance Colorless Liquid
Boiling Point 82°C
Quantity 1 L
Packaging Natural Poly Bottle
Specific Gravity 0.874 to 0.882
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Percent Purity 68 to 72% v/v
Grade Laboratory
Chemical Name or Material Isopropanol
Show More Show Less
ShelfLife 24 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.