Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol, 99.9%, for HPLC, ACROS Organics™

Click to view available options
Quantity:
4L
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
For HPLC
Absorbance (1cm cell vs water), 0.02 max. at 254nm, 0.13 max. at 230nm, 0.25 max. at 220nm, 1 max. at 205nm
Absorbance Conditions (1cm cell vs water)
Additional Information Vapor Pressure: 43mmHg at 20°C
Assay Percent Range 99.9%
Linear Formula (CH3)2CHOH
Beilstein 01, 360
Fieser 13,108
Merck Index 15, 5254
Formula Weight 60.1
Percent Purity ≥99.9%
Grade HPLC
Density 0.7850g/mL
Fluorescence Background Pass Test
Identification Pass Test
Refractive Index 1.3740 - 1.3760
Residue after Evaporation 2ppm max.
Solubility in Water Pass Test
Titratable Acid 0.0001mEq/g max.
Titratable Base 0.0001mEq/g max.
Water 0.1% max.
Flash Point 12°C (53.6°F)
Color Undesignated
Melting Point -89.5°C
Quantity 4L
Chemical Name or Material Isopropanol, 99.9%
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger