Learn More
Isopropanol/Water, 2:1 with Potassium Hydroxide, Reagents
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 88°C |
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.89 |
| Grade | Reagent |
| Chemical Name or Material | Isopropanol/Water, 2:1 with Potassium Hydroxide |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.