Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropenyl acetate, 99%

Catalog No. aaa15618ap
Click to view available options
Quantity:
500 mL

CAS: 108-22-5 | C5H8O2 | 100.12 g/mol

Iodine in isopropenyl acetate is a unique catalyst for the acetylation of a variety of alcohols, phenols and amines under solvent free conditions. It is used as a solvent for cellulose, plastics, oil and fats. It is also a building block used for various organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine in isopropenyl acetate is a unique catalyst for the acetylation of a variety of alcohols, phenols and amines under solvent free conditions. It is used as a solvent for cellulose, plastics, oil and fats. It is also a building block used for various organic synthesis.

Solubility
Slightly soluble in water. Soluble in alcohol and propylene glycol, acetone and ethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-22-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008709
InChI Key HETCEOQFVDFGSY-UHFFFAOYSA-N
Synonym isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate
PubChem CID 7916
IUPAC Name prop-1-en-2-yl acetate
SMILES CC(=C)OC(C)=O

Specifications

Melting Point -93°C
Density 0.917
Boiling Point 93°C to 94°C
Flash Point 18°C (64°F)
Refractive Index 1.401
Quantity 500 mL
UN Number UN2403
Beilstein 1280347
Merck Index 14,5203
Solubility Information Slightly soluble in water. Soluble in alcohol and propylene glycol,acetone and ethyl ether.
Formula Weight 100.12
Percent Purity 99%
Chemical Name or Material Isopropenyl acetate
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPENYL ACETATE
EINECSNumber 203-562-7
RTECSNumber UD4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.