Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropenyl Acetate 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
108-22-5
C5H8O2
100.12
MFCD00008709
HETCEOQFVDFGSY-UHFFFAOYSA-N
isopropenyl acetate,2-acetoxypropene,1-methylvinyl acetate,2-acetoxypropylene,methylvinyl acetate,1-propen-2-ol, acetate,propen-2-yl acetate,acetic acid isopropenyl ester,1-acetoxy-1-methylethylene,1-propen-2-yl acetate
7916
prop-1-en-2-yl acetate
CC(=C)OC(=O)C
108-22-5
C5H8O2
100.12
MFCD00008709
HETCEOQFVDFGSY-UHFFFAOYSA-N
isopropenyl acetate,2-acetoxypropene,1-methylvinyl acetate,2-acetoxypropylene,methylvinyl acetate,1-propen-2-ol, acetate,propen-2-yl acetate,acetic acid isopropenyl ester,1-acetoxy-1-methylethylene,1-propen-2-yl acetate
7916
prop-1-en-2-yl acetate
CC(=C)OC(=O)C
Melting Point -93°C
Boiling Point 94°C to 97°C
Flash Point 10°C
Specific Gravity 0.92
Refractive Index 1.4000 to 1.402
Quantity 1kg
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Density 0.9200g/mL
Assay Percent Range 99%
Linear Formula CH3CO2C(CH3)=CH2
Beilstein 02, II, 278
Fieser 01,524; 04,273; 06,308
Merck Index 15, 5249
Solubility Information Solubility in water: 34g/L in water (20°C). Other solubilities: miscible with nearly all organic solvents
Formula Weight 100.12
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Isopropenyl acetate, 99%
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-562-7

RTECSNumber : UD4200000

TSCA : TSCA