Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. p-3771107
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle

CAS: 13291-18-4 | C3H5BrMg | 145.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13291-18-4, 109-99-9
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00192240
InChI Key LFTYALBVGVNGLI-UHFFFAOYSA-M
Synonym isopropenylmagnesium bromide, isopropenylmagnesium bromide solution, isopropenyl magnesium bromide, bromo prop-1-en-2-yl magnesium, magnesium, bromo 1-methylethenyl, bromo isopropenyl magnesium, 1-methylethenylmagnesium bromide, 2-propen-2-ylmagnesium bromide, prop-1-en-2-ylmagnesium bromide, lftyalbvgvngli-uhfffaoysa-m, grignard reagent
PubChem CID 4187787
IUPAC Name magnesium;prop-1-ene;bromide
SMILES CC(=C)[Mg]Br

Specifications

Color Orange to White
Density 0.9270g/mL
Boiling Point 67.0°C
Flash Point 2°C
Packaging AcroSeal™ Glass bottle
Linear Formula CH2=C(CH3)MgBr
Quantity 100 mL
Specific Gravity 0.927
Formula Weight 145.28
Physical Form Crystalline Powder
Chemical Name or Material Isopropenylmagnesium bromide, 0.5M Solution in THF
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May form explosive
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.