Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IPA (Isopropyl Alcohol), 70% v/v, Fisher Chemical™ PROMO

Catalog No. A4594S24 Shop All Fisherbrand Products
Click to view available options
:
IPA (Isopropyl Alcohol); Packaging: Poly Bottle; Quantity: 4 L

Synonym: Isopropanol, 2-propanol, Propan-2-ol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0, 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -88°C
Color Colorless
Density 0.785 g/cm3
Boiling Point 82°C
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Refractive Index 1.3706 to 1.3722 (at 25°C)
Assay Percent Range 68 to 72%
Vapor Pressure 33 mmHg at 20°C
Formula Weight 60.1
Percent Purity 68 to 72%
Chemical Name or Material Isopropyl Alcohol
Show More Show Less
Hazard Statement Flammable liquid and vapor.
Precautionary Statement May cause respiratory irritation. May be harmful if inhaled.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.