Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Honeywell

Catalog No. p-7112052 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 gal.
19 L
4 x 4 L
6 x 1 L
204 L
Packaging:
Fiberboard Box
Glass Bottle
Metal Can
Metal Drum
Poly Bottle

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100.0000%
Density 0.785 g/cm3
Boiling Point 82.3°C
Quantity 6 x 1 L
Flash Point 12°C
Packaging Poly Bottle
UN Number UN1219
Beilstein 635639
Water Passes test
Solubility Information Miscible in Water at 20°C
Formula Weight 60.11
Percent Purity >99.5%
Grade ACS Reagent
Assay by GC Analysis
Chemical Name or Material Isopropyl Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.