Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, (Propanol; Isopropanol), Reagents

Catalog No. 01338115
Change view
Click to view available options
:
Quantity: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-338-115 Quantity: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-115 Supplier Reagents Holdings Llc Supplier No. C112155020E6
Only null left
Add to Cart
Add to Cart

Isopropyl Alcohol, ASTM D770 (2-Propanol; Isopropanol), ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Isopropanol, 2-Propanol, Propan-2-ol
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Color Colorless
Boiling Point 82.5°C
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.7808
Certifications/Compliance ASTM
Chemical Name or Material Iso-Propyl Alcohol, ASTM D770 (2-Propanol, Isopropanol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.