Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl-beta-D-thiogalactoside, High Purity, Spectrum™ Chemical
SDP

Catalog No. 18603581 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
6 x 1 g
6 x 5 g
100 g
1 kg
Packaging:
Amber Glass Bottle
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-581 25 g Amber Glass Bottle
18-603-575 100 mg Amber Glass Bottle
18-603-576 250 mg Amber Glass Bottle
18-603-577 1 g Amber Glass Bottle
18-603-579 5 g Amber Glass Bottle
18-603-578 6 x 1 g Amber Glass Bottle
18-603-580 6 x 5 g Amber Glass Bottle
18-603-582 100 g Amber Glass Bottle
18-603-583 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18-603-581 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I105525GM
Add to Cart
Add to Cart

I1055, 367-93-1, C9H18O5S

Spectrum™ Chemical Isopropyl-beta-D-thiogalactoside, High Purity (IPTG) is a compound and a molecular biology reagent. IPTG is an effective inducer of protein expression in the concentration range of 100 μM to 1.5 mM. Spectrum offers highly pure reagents suitable for molecular biology research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Grade High Purity
Physical Form Powder or Waxy Solid
Chemical Name or Material Isopropyl-beta-D-thiogalactoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.