Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isopropyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ A1164106

Quantity 5g

 View more versions of this product

Catalog No. AAA1164106


  Check Availability
Add to cart

Specifications

Description

Applications
Isopropyl isothiocyanate is used as is an antimicrobial agent. It is also used as medicinal intermediate.

Solubility
Miscible with methanol. Hydrolyzes in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Incompatible with alcohols and amines.
Description & Specifications

Chemical Identifiers

2253-73-8
101.17
UN2924
Isopropyl isothiocyanate
(CH3)2CHNCS
MFCD00037830
1739163

Specifications

40°C (104°F)
137° to 138°C
2253-73-8
101.17
UN2924
Isopropyl isothiocyanate
Pungent
5g
(CH3)2CHNCS
MFCD00037830
1739163
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit